Learn More
Biolegend Brilliant Violet 421™ anti-Tubulin Beta 3 (TUBB3)
Supplier:  Biolegend 657412
Encompass_Preferred
Brilliant Violet 421™ anti-Tubulin Beta 3 (TUBB3) [AA10]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Rat, Human; Apps: ICC, IHC-F; Size: 100 ug
Catalog No. 50-114-5231