Learn More
Biolegend Brilliant Violet 421™ anti-DYKDDDDK Tag
Catalog No. 502086866
Encompass_Preferred
Brilliant Violet 421™ anti-DYKDDDDK Tag [L5]; Isotype: Rat IgG2a, λ; Reactivity: DYKDDDDK tag epitope; Apps: FC; Size: 25 ug
Catalog No. 50-208-6866 Supplier Biolegend Supplier No. 637321
Brilliant Violet 421™ anti-DYKDDDDK Tag [L5]; Isotype: Rat IgG2a, λ; Reactivity: DYKDDDDK tag epitope; Apps: FC; Size: 25 ug