Learn More
Biolegend Brilliant Violet 421™ anti-GATA3
Supplier:  Biolegend 653813
Encompass_Preferred
Brilliant Violet 421™ anti-GATA3 [16E10A23]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-712-572