Learn More
Biolegend Brilliant Violet 421™ anti-human CD11c
Supplier:  Biolegend 301627
Encompass_Preferred
Brilliant Violet 421™ anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. 50-165-733