Learn More
Biolegend Brilliant Violet 421™ anti-human CD19
Supplier:  Biolegend 302234
Encompass_Preferred
Brilliant Violet 421™ anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC, ICC, IHC-F; Size: 100 tests
Catalog No. 50-403-206