Learn More
Biolegend Brilliant Violet 421™ anti-human CD192 (CCR2)
Supplier:  Biolegend 357210
Encompass_Preferred
Brilliant Violet 421™ anti-human CD192 (CCR2) [K036C2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-156