Learn More
Biolegend Brilliant Violet 421™ anti-human CD200 (OX2)
Supplier:  Biolegend 329209
Encompass_Preferred
Brilliant Violet 421™ anti-human CD200 (OX2) [OX-104]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 25 tests
Catalog No. 50-207-5841