Learn More
Biolegend Brilliant Violet 421™ anti-human CD200R
Supplier:  Biolegend 329313
Encompass_Preferred
Brilliant Violet 421™ anti-human CD200R [OX-108]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC; Size: 25 tests
Catalog No. 50-207-5861