Learn More
Biolegend Brilliant Violet 421™ anti-human CD326 (Ep-CAM)
Supplier:  Biolegend 324219
Encompass_Preferred
Brilliant Violet 421™ anti-human CD326 (Ep-CAM) [9C4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-239