Learn More
Biolegend Brilliant Violet 421™ anti-human CD34
Supplier:  Biolegend 343609
Encompass_Preferred
Brilliant Violet 421™ anti-human CD34 [561]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-403-368