Learn More
Biolegend Brilliant Violet 421™ anti-human CD4
Supplier:  Biolegend 317434
Encompass_Preferred
Brilliant Violet 421™ anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC, ICC; Size: 100 tests
Catalog No. 50-604-134