Learn More
Biolegend Brilliant Violet 421™ anti-human CD49d
Supplier:  Biolegend 304322
Encompass_Preferred
Brilliant Violet 421™ anti-human CD49d [9F10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cynomolgus, Dog (Canine), Horse (Equine), Rhesus, Sheep (Ovine), Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-850-538