Learn More
Biolegend Brilliant Violet 421™ anti-human CD73 (Ecto-5'-nucleotidase)
Supplier:  Biolegend 344008
Encompass_Preferred
Brilliant Violet 421™ anti-human CD73 (Ecto-5'-nucleotidase) [AD2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC, IHC-F; Size: 100 tests
Catalog No. 50-403-370