Learn More
Biolegend Brilliant Violet 421™ anti-human CD80
Supplier:  Biolegend 305222
Encompass_Preferred
Brilliant Violet 421™ anti-human CD80 [2D10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-850-547