Learn More
Biolegend Brilliant Violet 421™ anti-human CD95 (Fas)
Supplier:  Biolegend 305624
Encompass_Preferred
Brilliant Violet 421™ anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests
Catalog No. 50-604-056