Learn More
Biolegend Brilliant Violet 421™ anti-human CD96 (TACTILE)
Supplier:  Biolegend 338418
Encompass_Preferred
Brilliant Violet 421™ anti-human CD96 (TACTILE) [NK92.39]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-112-9871