Learn More
Biolegend Brilliant Violet 421™ anti-human IFN-γ
Supplier:  Biolegend 502531
Encompass_Preferred
Brilliant Violet 421™ anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ICFC; Size: 25 tests
Catalog No. 50-169-751