Learn More
Biolegend Brilliant Violet 421™ anti-human IL-13
Supplier:  Biolegend 501915
Encompass_Preferred
Brilliant Violet 421™ anti-human IL-13 [JES10-5A2]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-5535