Learn More
Biolegend Brilliant Violet 421™ anti-human IL-17A
Supplier:  Biolegend 512322
Encompass_Preferred
Brilliant Violet 421™ anti-human IL-17A [BL168]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-403-676