Learn More
Biolegend Brilliant Violet 421™ anti-human IL-4
Supplier:  Biolegend 500826
Encompass_Preferred
Brilliant Violet 421™ anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ICFC; Size: 100 tests
Catalog No. 50-604-728