Learn More
Biolegend Brilliant Violet 421™ anti-human TCR Vα7.2
Supplier:  Biolegend 351716
Encompass_Preferred
Brilliant Violet 421™ anti-human TCR Vα7.2 [3C10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-103-0776