Learn More
Biolegend Brilliant Violet 421™ anti-human CD11c
Supplier:  Biolegend 371512
Encompass_Preferred
Brilliant Violet 421™ anti-human CD11c [S-HCL-3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8658