Learn More
Biolegend Brilliant Violet 421™ anti-human CD122 (IL-2Rβ)
Supplier:  Biolegend 339010
Encompass_Preferred
Brilliant Violet 421™ anti-human CD122 (IL-2Rβ) [TU27]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1575647