Learn More
Biolegend Brilliant Violet 421™ anti-human CD19
Supplier:  Biolegend 363017
Encompass_Preferred
Brilliant Violet 421™ anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-F; Size: 25 tests
Catalog No. 50-207-8115