Learn More
Biolegend Brilliant Violet 421™ anti-human CD38
Supplier:  Biolegend 356618
Encompass_Preferred
Brilliant Violet 421™ anti-human CD38 [HB-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-7570