Learn More
Biolegend Brilliant Violet 421™ anti-human CD62P (P-Selectin)
Supplier:  Biolegend 304926
Encompass_Preferred
Brilliant Violet 421™ anti-human CD62P (P-Selectin) [AK4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1207731