Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD163
Supplier:  Biolegend 155309
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD163 [S15049I]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1713444