Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD304 (Neuropilin-1)
Supplier:  Biolegend 145209
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD304 (Neuropilin-1) [3E12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-605-999