Learn More
Biolegend Brilliant Violet 421™ anti-mouse IL-33Rα (IL1RL1, ST2)
Supplier:  Biolegend 145309
Encompass_Preferred
Brilliant Violet 421™ anti-mouse IL-33Rα (IL1RL1, ST2) [DIH9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-207-1615