Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD117 (c-kit)
Supplier:  Biolegend 105828
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD117 (c-kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 uL
Catalog No. 50-402-972