Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD11c
Supplier:  Biolegend 117330
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC; Size: 500 uL
Catalog No. 50-403-003