Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD138 (Syndecan-1)
Supplier:  Biolegend 142523
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD138 (Syndecan-1) [281-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-103-0566