Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD152
Supplier:  Biolegend 106312
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-604-865