Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD170 (Siglec-F)
Supplier:  Biolegend 155509
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD170 (Siglec-F) [S17007L]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-207-2208