Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD183 (CXCR3)
Supplier:  Biolegend 126521
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-164-309