Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD19
Supplier:  Biolegend 115549
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 50 ug
Catalog No. 50-604-902