Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD197 (CCR7)
Supplier:  Biolegend 120119
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-163-837