Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD206 (MMR)
Supplier:  Biolegend 141717
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC, IHC-F; Size: 125 uL
Catalog No. 50-605-994