Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD3ε
Supplier:  Biolegend 100341
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, ICC; Size: 50 ug
Catalog No. 50-604-778