Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD4
Supplier:  Biolegend 100438
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, ICC, IHC-F; Size: 500 uL
Catalog No. 50-402-919