Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD45.1
Supplier:  Biolegend 110731
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-163-434