Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD68
Supplier:  Biolegend 137017
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD68 [FA-11]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: ICFC, FC; Size: 50 ug
Catalog No. 50-605-040