Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD80
Supplier:  Biolegend 104726
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC, IHC-F; Size: 500 uL
Catalog No. 50-402-967