Learn More
Biolegend Brilliant Violet 421™ anti-mouse F4/80
Supplier:  Biolegend 123132
Encompass_Preferred
Brilliant Violet 421™ anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 500 uL
Catalog No. 50-403-013