Learn More
Biolegend Brilliant Violet 421™ anti-mouse IL-9
Supplier:  Biolegend 514109
Encompass_Preferred
Brilliant Violet 421™ anti-mouse IL-9 [RM9A4]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 ug
Catalog No. 50-208-5871