Learn More
Biolegend Brilliant Violet 421™ anti-mouse LAP (TGF-β1)
Supplier:  Biolegend 141407
Encompass_Preferred
Brilliant Violet 421™ anti-mouse LAP (TGF-β1) [TW7-16B4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-403-070