Learn More
Biolegend Brilliant Violet 421™ anti-mouse Ly-6C
Supplier:  Biolegend 128032
Encompass_Preferred
Brilliant Violet 421™ anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 500 uL
Catalog No. 50-605-983