Learn More
Biolegend Brilliant Violet 421™ anti-mouse/rat XCR1
Supplier:  Biolegend 148216
Encompass_Preferred
Brilliant Violet 421™ anti-mouse/rat XCR1 [ZET]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse, Rat; Apps: FC; Size: 50 ug
Catalog No. 50-103-0583