Learn More
Biolegend Brilliant Violet 421™ anti-mouse TNF-α
Supplier:  Biolegend 506328
Encompass_Preferred
Brilliant Violet 421™ anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 ug
Catalog No. 50-605-489