Learn More
Biolegend Brilliant Violet 421™ anti-STAT3 Phospho (Tyr705)
Supplier:  Biolegend 651009
Encompass_Preferred
Brilliant Violet 421™ anti-STAT3 Phospho (Tyr705) [13A3-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-6998